Learn More
Ácido piperonílico, +98 %, Thermo Scientific Chemicals
Catalog Number p-7021676
Click to view available options
Cantidad:
25 g
100 g
500 g
94-53-1
C8H6O4
166.13
MFCD00005830
VDVJGIYXDVPQLP-UHFFFAOYSA-N
piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid
7196
2H-1,3-benzodioxole-5-carboxylic acid
OC(=O)C1=CC=C2OCOC2=C1
94-53-1
C8H6O4
166.13
MFCD00005830
VDVJGIYXDVPQLP-UHFFFAOYSA-N
piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid
7196
2H-1,3-benzodioxole-5-carboxylic acid
OC(=O)C1=CC=C2OCOC2=C1
Piperonylic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El ácido piperonílico se utiliza como intermediario farmacéutico.
Solubilidad
Ligeramente soluble en agua.
Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Piperonylic acid
Punto de fusión 230°C to 235°C
Cantidad 25 g
Beilstein 150206
Índice Merck 14,7477
Información de solubilidad Slightly soluble in water.
Formula Weight (peso de la fórmula) 166.13
Porcentaje de pureza ≥98%