Learn More
3M™ Particulate Filters
Marca: 3M™ 7000029734
Detalles adicionales : Peso : 0.80000kg
Especificaciones
P3 R | |
Pharmaceutical, powered chemicals, construction, quarrying ceramics, and refractory materials | |
2135 |
Particulate Filter | |
Suitable for 3M™ 6000 and 3M™ 7000 Series Half Face and Full Face Masks | |
20/Pk. |