Learn More
Thermo Scientific™ Owl™ Leveling Platform
Accommodate most applications with this three-point leveling platform which ensures a flat casting and running surface.
Marca: Thermo Scientific™ BBL-1
16.00 EUR hasta el 2024-05-31
Use el código promocional "21664" para aplicar la oferta.
Detalles adicionales : Peso : 2.00000kg
Descripción
Accommodate most applications with the Thermo Scientific™ Owl™ Leveling Platform, a three-point leveling platform that ensures a flat casting and running surface.
- Tamaño: 36 x 46cm
- Incluye nivel de burbuja
Especificaciones
Sistema Owl™ | |
Owl™ | |
Nivel de burbuja |