Learn More
Hughes Outdoor Self-Draining Safety Shower with Eye/Face Wash
This ANSI compliant self-draining emergency safety shower with open stainless steel eye/face wash, suitable for hot climates.
1127.69€ - 1617.40€
Especificaciones
Activación | Palanca de empuje, pedal |
---|---|
Tipo de producto | Ducha de seguridad |
Código de producto | Marca | Material | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Material | Precio | Cantidad y disponibilidad | |||||
17722348
|
Hughes
HSHDSD18GV |
Acero galvanizado |
1127.69€
cada uno |
Fecha estimada de envío: 30-05-2024
Inicie sesión para ver el stock |
|||||
17732348
|
Hughes
HSHDSD18SS |
Acero inoxidable |
1617.40€
cada uno |
Fecha estimada de envío: 30-05-2024
Inicie sesión para ver el stock |
|||||
Descripción
- This ANSI compliant self-draining emergency safety shower with open stainless steel eye/face wash, suitable for hot climates
- The design allows standing water to drain away from the standpipe to ensure the delivery of tepid water
- Fitted with foot panel as standard
- The valve is situated directly above the water inlet; the bleed pipe is fitted above this so standing water can drain into the ground preventing stagnant water overheating from solar radiation and removing the risk of further injury to the operator
- Available with galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Especificaciones
Palanca de empuje, pedal | |
Ducha de seguridad |