missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NCBP1 Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP1610
Detalles adicionales : Peso : 0.01000kg
Descripción
NCBP1 Synthetic Peptide, PEP-1610, from Invitrogen.
This peptide corresponds to 17 amino acids near the center of human NCBP1.PEP-1610 can be used as a blocking peptide with polyclonal antibody PA5-34567. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37ºC.Especificaciones
4686 | |
purified | |
Human NCBP1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q09161 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
NCBP1 | |
0.02% sodium azide | |
50 μg | |
PBS |