missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-propargil-D-glicina, 95 %, Thermo Scientific™

Catalog Number p-7041556
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
63039-46-3
C10H14NO4
212.23
MFCD01320886
AMKHAJIFPHJYMH-SSDOTTSWSA-M
boc-d-propargylglycine, r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, n-boc-2-propargyl-d-glycine, boc-d-pra-oh, s-n-boc-propargylglycine, 2r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-d-gly propargyl-oh, boc-r-2-propargylglycine, n-tert-butoxycarbonyl-3-ethynyl-d-alanine, 2-r-tert-butoxycarbonylaminopent-4-ynoic acid
7009126
CC(C)(C)OC(=O)N[C@H](CC#C)C([O-])=O
Este artículo no se puede devolver. Vea la política de devoluciones
63039-46-3
C10H14NO4
212.23
MFCD01320886
AMKHAJIFPHJYMH-SSDOTTSWSA-M
boc-d-propargylglycine, r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, n-boc-2-propargyl-d-glycine, boc-d-pra-oh, s-n-boc-propargylglycine, 2r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-d-gly propargyl-oh, boc-r-2-propargylglycine, n-tert-butoxycarbonyl-3-ethynyl-d-alanine, 2-r-tert-butoxycarbonylaminopent-4-ynoic acid
7009126
CC(C)(C)OC(=O)N[C@H](CC#C)C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Boc-2-propargyl-D-glycine
Cantidad 250 mg
Nombre IUPAC (2R)-2-{[(terc-butoxi)carbonil]amino}pent-4-inoato
Formula Weight (peso de la fórmula) 213.23
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'