missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS27A, Mouse, Clone: Ubi-1, Abnova™
Mouse monoclonal antibody raised against RPS27A.
Marca: Abnova MAB2414.100uL
Detalles adicionales : Peso : 0.02000kg
Especificaciones
RPS27A | |
Monoclonal | |
Unconjugated | |
Immunofluorescence (1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μL | |
Primary | |
Bovine, Chicken, Drosophila, Human, Mouse, Nematode, Plant | |
Liquid |
ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
Ubi-1 | |
Mouse monoclonal antibody raised against RPS27A. | |
In buffer containing 5mM sodium azide | |
Purified ubiquitin conjugated with KLH with glutaraldehyde. | |
RUO | |
This antibody specifically recognizes ubiquitinated cytoplasmic inclusion bodies. | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |