Learn More
GST, Mouse, Clone: 3D4, Abnova™
Mouse monoclonal antibody raised against GST.
Marca:  Abnova MAB1996.500ug
Detalles adicionales : Peso : 0.02000kg
Código de producto. 16055396
  • 730.00€ / 500 microgramos
Fecha estimada de envío: 11-06-2024
para ver el stock
Especificaciones
GST
Monoclonal
Unconjugated
ELISA (1:20000-1:100000) Western Blot (1:1000-1:10000) The optimal working dilution should be determined by the end user.
Mouse
500 μg
Primary
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG1 κ
ELISA, Western Blot
3D4
Mouse monoclonal antibody raised against GST.
In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Glutathione-S-Transferase (GST) from Schistosoma japonicum.
RUO
S. japonicum
Liquid
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only