missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aflatoxin B1, Mouse, Clone: 6A10, Abnova™
Mouse monoclonal antibody raised against Aflatoxin B1.
Marca: Abnova MAB2406.100uL
Detalles adicionales : Peso : 0.02000kg
Especificaciones
Aflatoxin B1 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μL | |
Primary | |
Fungi | |
Liquid |
ELISA, Immunofluorescence, Immunohistochemistry | |
6A10 | |
Mouse monoclonal antibody raised against Aflatoxin B1. | |
In PBS (0.09% sodium azide) | |
Aflatoxin B1 (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1). | |
RUO | |
This antibody is specific to Aflatoxin B1. | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |