Learn More
Motic™ Moticam A8
The fast sCMOS imaging setup of the Moticam A8 allows 4K resolution while delivering steady 15 fps. Printing images requires high-resolution data; Moticam delivers the detail needed.
Marca: Motic™ 1100600101443
Detalles adicionales : Peso : 0.41000kg
Descripción
The ideal camera, for everyone who has high demands and wants to stay flexible.
- C-mount for lens
- MotiConnect software is available for free download (with free updates included)
- Live Display Modes: 3840 x 2160 px, 1920 x 1080 px
- Maximum Frame Rate: 3840 x 2160 at 15 fps, 1920 x 1080 at 30 fps
Especificaciones
Camera | |
Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap | |
1/3 in. |
Moticam A | |
sCMOS | |
15 to 30 fps |