missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MOSPD1 Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP1572
Detalles adicionales : Peso : 0.01000kg
Descripción
MOSPD1 Synthetic Peptide, PEP-1572, from Invitrogen.
This peptide corresponds to 19 amino acids near the center of human MOSPD1.PEP-1572 can be used as a blocking peptide with polyclonal antibody PA5-34529. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37ºC.Especificaciones
56180 | |
purified | |
Human MOSPD1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9UJG1 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
MOSPD1 | |
0.02% sodium azide | |
50 μg | |
PBS |