Learn More
Invitrogen™ Molecular Probes™ Guanosine 5'-Triphosphate, BODIPY™ TR 2'-(or-3')-O-(N-(2-Aminoethyl) Urethane), Trisodium Salt (BODIPY TR GTP)
Exhibits spectral characteristics similar to the Texas Red™ dye
Marca: Invitrogen™ G22351
Detalles adicionales : Peso : 0.00450kg
Descripción
The mixed-isomer analog of BODIPY TR GTP is comprised of the BODIPY TR fluorophore attached to the 2' or 3' position of the ribose ring via an aminoethylcarbamoyl linker. BODIPY TR GTP is significantly quenched in solution and increase upon binding to at least some GTP-binding proteins. BODIPY TR exhibits spectral characteristics similar to the Texas Red dye. This fluorescent nucleotide may be useful for fluorescence polarization-based assays of GTP-binding proteins.
Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation
Información de órdenes
Shipping Condition: Wet ice
Especificaciones
BODIPY TR | |
100 μL | |
Store in freezer (-5°C to -30°C) and protect from light. |
Ribonucleotide, BODIPY dye modified | |
5mM |
For Research Use Only. Not for use in diagnostic procedures.