missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABgene™ Microplate, Thermo-Fast PCR/qPCR, 96 well, nonskirted, standard, profile

Marca:  Thermo Scientific™ ABgene™ AB0700 - RMD142

Detalles adicionales : Peso : 0.50000kg

Código de producto. 15699504

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »


Descripción

Descripción

  • 0.3mL maximum well volume (when used with adhesive and heat seals)
  • Fits all 0.2mL thermal cyclers and ABI PRISM™ 310, 3100 and 3700 capillary sequencers
  • Clear well bottom for sample visibility
  • Highly rigid plate deck, with cut away corners
  • Sealable with Thermo-Mats™, adhesive films and foils, heat seals and flat and domed cap strips
  • Available in packs of 25
  • Also available in multiple colours and opaque black and white for fluorescent and luminescent based procedures
  • Uniform, thin wall design for optimal heat transfer
  • Certified DNase, RNase, endotoxin and human DNA freeA one piece, injection moulded polypropylene PCR* plate in the industry standard 96 well format, providing optimal handling convenience and compatibility with high throughput automated systems
Especificaciones

Especificaciones

Standard
8 x 12
96
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales