missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microglass Fiber Filters, Grade 227
Suitable for micro-filtration and can be used in various fields.
Marca: Fisherbrand™
Detalles adicionales : Peso : 0.01000kg
Descripción
- High level of particle retension
- Large load capacity
- High chemical purity (made with demineralized water)
- Non-hygroscopic and biologically inert
- High permeability to air
- Chemical resistance to most solvents and reagents (except hydrofluoric acid)
- Resist to temperature of about 500°C
Especificaciones
70 g/m2 | |
Disc | |
0.7 to 2.7 μm | |
227 | |
500°C | |
0.4 mm | |
70 mm |
Microfilter | |
Borosilicate Glass | |
Air pollution control, separation of soot before gas analyzers, support weighing in the balance analyzes and dryers | |
Non-hygroscopic | |
0.015 in. | |
Filter |