missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabolite Benzo(g,h,i)perylene in DCM 100μg/mL, Fisher Chemical™
Click to view available options
Cantidad:
1 mL
Envase:
Ampoule(s)
Especificaciones
Nombre del producto químico o material | Benzo(g,h,i)perylene |
Concentración | 100μg/mL |
CAS | 191-24-2 |
Envase | Ampoule(s) |
Cantidad | 1 mL |
Fórmula molecular | C22H12 |
Clave InChI | GYFAGKUZYNFMBN-UHFFFAOYSA-N |
SMILES | C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2 |
Peso molecular (g/mol) | 276.338 |
PubChem CID | 9117 |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'