missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(Ii) Sulfate, Honeywell Fluka™
Click to view available options
Cantidad:
250 g
1 kg
6 x 250 g
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Especificaciones
Nombre del producto químico o material | Mercury(Ii) Sulfate |
Cantidad | 1 kg |
Fórmula lineal | HgSO4 |
Número UN | 1645 |
Nombre IUPAC | mercury(2+);sulfate |
Formula Weight (peso de la fórmula) | 296.65g/mol |
Porcentaje de pureza | ≥98% |
Grado | puriss. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'