Learn More
MAP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA585755
Detalles adicionales : Peso : 0.01000kg
Descripción
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%). Positive Control: mouse brain, DIV9 rat E18 primary cortical neurons. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's Disease (AD).Especificaciones
MAP2 | |
Polyclonal | |
Unconjugated | |
Map2 | |
a730034c02, AC091465.1, DKFZp686I2148, G1-397-34, MAP 2, Map2, MAP-2, map2.S, map2a, map-2a, map2ab, MAP2B, MAP2C, MAP2R, microtubule associated protein 2, microtubule associated protein 2 S homeolog, microtubule associated protein 2, microtubule-associated protein 2, microtubule associated protein 2C, microtubule-associated protein 2, microtubule-associated protein-2, Mtap2, Mtap-2, OTTHUMP00000163916, OTTHUMP00000208143, repro4, XELAEV_18047318mg, xmap2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17756, 25595, 4133 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.07 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300, pH 7 | |
P11137, P15146, P20357 | |
Map2 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Mouse MAP2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |