missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Advantec Mfs™ Soporte de filtro a presión KST 90
KST 90 Pressure Filter Holder with reservoir is designed for ultra cleaning and sterilizing small batch volumes of liquids without the need for a separate dispensing reservoir or pressure valve.
Marca: Advantec Mfs™ 361600
Detalles adicionales : Peso : 0.80000kg
Descripción
- Available in both 304 or 316 stainless steel
- PTFE coated support screen permits autoclaving with the filter in place
- Unit is easily and quickly assembled, simple hand tightening of the wing-nuts achieves an integral seal
- Separate ports are used to pressurize the reservoir and for adding the sample
- Suitable for hazardous toxicity testing and TCLP
Especificaciones
Autoclavable | |
Pressure filter holder with reservoir | |
Ultra cleaning buffers; sterilizing small batches of media or serum; flow decay studies in pilot production operations; suitable for hazardous toxicity testing and TCLP | |
Adaptadores de manguera dentada y válvula de escape de presión | |
750 mL |
NPT | |
Stainless Steel (304) | |
304 Stainless steel | |
Paquete de 1 | |
90 mm |