Learn More
DWK Life Sciences Kimble™ Brand Rotary Evaporator: Bottom Vapor Tube Traps
Accessory for rotary evaporator trap
Marca: DWK Life Sciences 5702652524
Detalles adicionales : Peso : 0.80000kg
Descripción
- Square Joint Bead
Especificaciones
4mm drain holes | |
Bottom Vapor Tube Trap | |
Used when foam is likely to be created and drawn into the condenser of rotary evaporator | |
Upper: 24/40, Lower: 24/40 | |
Borosilicate Glass | |
USP Type I and ASTM E438, Class A |
250 mL | |
8.45 oz., 250 mL | |
20.5 cm | |
8 in. | |
8.45 oz. |