Learn More
3M™ Ioban™ 2 Antimicrobial Incise Drape
3M™ Ioban™ 2 Antimicrobial Incise Drape provides a sterile surface to the wound edge at the start of surgery and continuous antimicrobial activity throughout the procedure.
Marca: 3M™ 7000042935
Detalles adicionales : Peso : 0.70000kg
Descripción
- Superior adhesion
- Excellent antimicrobial
- Designed for infection prevention of surgical sites
Especificaciones
60 x 45 cm | |
Surgery | |
Sterile |
Incise Drapes | |
45 cm | |
60 cm |