missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Versilon™ Chloroprene Flexible Tubing

Versilon™ Chloroprene Flexible Tubing is heat resistant for industrial applications.

Marca:  Saint Gobain Life Sciences 720345

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15670181

  • 155.00€ / paquete de 50
Fecha estimada de envío: 16-07-2024
para ver el stock
Añadir a la cesta

Descripción

Descripción

With its good mechanical, chemical, electrical, and heat resistant (thermal) properties, Versilon™ Chloroprene Flexible Tubing from Saint-Gobain is recommended for a wide range of industrial applications. Its economic cost makes it an ideal industrial tubing. Chloroprene flexible tubing is used as much for cable insulation as for transfer of nitrogen and helium gases. It is recommended for welding equipment, automotive, and electro-mechanical industries. Additionally, chloroprene tubing shows an outstanding resistance to UV and weathering.

  • Resistance to hydrocarbons and mineral oils
  • Good ageing; ozone resistant
  • High dielectrical strength

Typical Applications

  • General industrial
  • Cable insulation
  • Nitrogen and helium gas transfer
  • Welding equipment
  • Automotive
  • Electro-mechanical industries
Especificaciones

Especificaciones

Tubing
5 mm
8 mm
1 m
1.5 mm
-20 °C
60 lb-f/in
Negro
Los tubos
de 100 °C
0.197 in.
Caucho sintético
164 ft.
0.06 in.
-4°F
50 m
Non-sterile
50 (Shore A)
212°F
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales