missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iduronate 2 Sulfatase Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA547331
Detalles adicionales : Peso : 0.00010kg
Descripción
In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human IDS is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Iduronate 2 Sulfatase Polyclonal antibody specifically detects Iduronate 2 Sulfatase in Mouse samples. It is validated for Immunoprecipitation, Western Blot.Especificaciones
Iduronate 2 Sulfatase | |
Polyclonal | |
Unconjugated | |
Ids | |
alpha-L-iduronate sulfate sulfatase; AW214631; Ids; Iduronate 2-sulfatase; iduronate 2-sulfatase (Hunter syndrome); iduronate 2-sulfatase 14 kDa chain; Iduronate 2-sulfatase 42 kDa chain; iduronate sulfatase; iduronate-2-sulfatase; idursulfase; MPS2; SIDS | |
Goat | |
Antigen affinity chromatography | |
RUO | |
15931 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q08890 | |
Ids | |
Mouse myeloma cell line NS0-derived recombinant mouse Iduronate 2-Sulfatase/IDS Thr36-Pro552 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |