missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iduronate 2 Sulfatase Monoclonal Antibody (OTI4G2), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA525866
Detalles adicionales : Peso : 0.01000kg
Descripción
Iduronate-2-sulfatase is required for the lysosomal degradation of heparan sulfate and dermatan sulfate. Mutations in this X-chromosome gene that result in enzymatic deficiency lead to the sex-linked Mucopolysaccharidosis Type II, also known as Hunter Syndrome. Iduronate-2-sulfatase has a strong sequence similarity with human arylsulfatases A, B, and C, and human glucosamine-6-sulfatase. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described.Especificaciones
Iduronate 2 Sulfatase | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22304, Q08890 | |
Ids | |
Full length human recombinant protein of IDS produced in HEK293T cell | |
100μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4G2 | |
Unconjugated | |
Ids | |
alpha-L-iduronate sulfate sulfatase; AW214631; Ids; Iduronate 2-sulfatase; iduronate 2-sulfatase (Hunter syndrome); iduronate 2-sulfatase 14 kDa chain; Iduronate 2-sulfatase 42 kDa chain; iduronate sulfatase; iduronate-2-sulfatase; idursulfase; MPS2; SIDS | |
Mouse | |
Affinity Chromatography | |
RUO | |
15931, 3423, 363513, 492194 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |