missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cloruro de hidrógeno, de 5 a 6 M en 2-propanol, puro, Thermo Scientific Chemicals
Click to view available options
Cantidad:
100 mL
500 mL
Descripción
Hydrogen chloride, 5 to 6M soln. in 2-propanol may be used in measuring their conductance at 250. It is used in the manufacturing of pharmaceuticals to get a hydrochloride salt. For example, ranitidine base is converted to ranitidine hydrochloride, which is used as anti ulcerative drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Hydrochloric acid |
Notas de grado de pureza | Pure |
Fórmula lineal | HCl |
Número UN | UN2924 |
Concentración | 5 to 6M soln. in 2-propanol |
Densidad | 0.909 |
Información de solubilidad | Miscible with water,methanol,ethanol and 2-propanol. |
Nombre IUPAC | hydrogen chloride |
Formula Weight (peso de la fórmula) | 36.46 |
Olor | Alcohol-like |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'