Learn More
HCCS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551783
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: YVECPIRGTA AENKENLDPS NLMPPPNQTP APDQPFALST VREESSIPRA DSEKKWVYPS EQMFWNAMLK KGWKWKDEDI SQKDMYNIIR IHNQNNEQAW KEILKWEALH AAECPCGPSL IRFGGKAKEY SPRARIRSWM GYEL Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
HCCS Polyclonal antibody specifically detects HCCS in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
HCCS | |
Polyclonal | |
Unconjugated | |
HCCS | |
CCHL; cytochrome c heme-lyase; cytochrome c-type heme lyase; HCCS; holocytochrome c synthase; holocytochrome c synthetase; holocytochrome c-type synthase; LSDMCA1; MCOPS7; microphthalamia with linear skin defects; MLS; RGD1563855 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15159, 3052, 317444 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P53701, P53702 | |
HCCS | |
Recombinant protein corresponding to Human HCCS | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only