missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Embudo de filtración de vidrio de repuesto de vidrio borosilicato GV050
This Glass Filter Funnel is a replacement part for Whatman™ GV 050 Series Vacuum Filtration Manifolds.
Marca: Cytiva 10442001
Detalles adicionales : Peso : 0.26320kg
Descripción
- Description: Replacement Part for GV 050 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Especificaciones
Autoclavable | |
Reusable | |
Vidrio de borosilicato | |
Vacuum Filtration | |
1 unidad | |
Filter Funnel |
Filter Funnel (Upper Section) | |
Para soportes de filtro de vacío de vidrio de la serie GV050 | |
Non-sterile | |
GV050/0/01 | |
GV050 | |
250 mL |