missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR91 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA598179
Detalles adicionales : Peso : 0.01000kg
Descripción
This polyclonal antibody was raised in Rabbit against GPR91. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPR91. BLAST analysis of the peptide immunogen showed no homology with other human proteins.Especificaciones
GPR91 | |
Polyclonal | |
Unconjugated | |
SUCNR1 | |
G protein-coupled receptor 91, GPR91, G-protein coupled receptor 91, G-protein-coupled receptor 91, p2u2, P2Y purinoceptor 1, P2Y purinoceptor 1-like, Succinate receptor, Succinate receptor 1, SUCNR1, SUCR1 | |
Rabbit | |
Protein G | |
RUO | |
56670 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4 | |
Q9BXA5 | |
SUCNR1 | |
Recombinant Human Succinate receptor 1 protein (303-334AA) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |