Learn More
Goat anti-Human IgG F(ab')2 Cross-Adsorbed Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Marca: Invitrogen 31132
Detalles adicionales : Peso : 0.80000kg
Descripción
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31132 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31132 reacts with the light chains of human IgG and with the light chains common to other human immunoglobulins, but not with the Fc portion of human IgG or non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, horse and mouse serum proteins. However, the antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of origin: USA
Human IgG F(ab')2 Cross-Adsorbed Polyclonal antibody specifically detects Human IgG F(ab')2 Cross-Adsorbed in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Especificaciones
Human IgG F(ab')2 Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Goat | |
1.5 mg | |
Secondary | |
4° C | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1.83 mg/mL | |
PBS with no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Human | |
Liquid |