Learn More
Gibco™ DMEM w/High Glucose and w/o Glutamine, Lysine and Arginine
Used during SILAC™ protein labeling with stable isotopic labeled lysine and/or arginine
Marca: Gibco A1443101
Detalles adicionales : Peso : 0.61000kg
Descripción
Certifications
ISO 13485