Learn More
BTX™ Flat Tweezertrode Platinum Kit
Flat Tweezertrode Platinum is a reusable non-invasive, tweezer-style electrodes for drug or gene delivery in animal tissues.
Marca: BTX™ 450525
Detalles adicionales : Peso : 0.40000kg
Descripción
Applications:
- In vivo applications
- In utero and ex-vivo gene transfection
- Eletroporation therapy
- Electropchemotherapy
- Transdermal drug delivery
Compatability:
- ECM830
- ECM2001+
- Gemini X2
- Connect to BTX generators using the Tweezertrode Connection cables (45-0204)
Tweezertrodes may be cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide.
Especificaciones
Not Autoclavable | |
ECM 830, ECM 2001, Gemini X2 | |
Cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide. | |
0.04 in. |
Reusable | |
Non-sterile | |
5°C to 40°C | |
1 mm |