missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ DVB/Sulfate Latex Beads, 4% w/v, 0.1μm

DVB/Sulfate Latex Beads, 4% w/v, 0.1 μm

Marca:  Molecular Probes™ S37391

Detalles adicionales : Peso : 0.00450kg

Código de producto. 11590696

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

The DVB/sulfate microspheres are negatively charged cross-linked polystyrene microspheres with sulfate functional groups on the surface. The surface is hydrophobic in nature.

Microspheres Are Available in Many Sizes and Surface Functionalities
  • 25 sizes to choose from—0.02 microns to 16 microns
  • 20 different surface—for ready conjugation to virtually any material
  • Surfactant free—ultraclean
  • Various quantities—from milliliter to 500-liter amounts

    Key Applications of Microspheres
  • Instrument calibration (flow cytometry, microscopy, HTS, HCS)
  • Flow testing (microfluidics, blood flow, water flow, and air flow)
  • Cell biology tracers (cell differentiation and cell tracing)
  • Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

    Get More Information
    Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

    We’ll Make a Custom Microsphere Product for You
    If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.

    For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
  • Especificaciones

    Especificaciones

    0.04w/v
    DVB/Sulfate Latex Beads, 15mL, 4% w/v Concentration, 0.1μm Particle Size, White, Suspension Form, De-ionized Water Medium, Specialty Latex Beads, Room Temperature, 2 to 8°C Storage Temperature, Ideal Materials for the Adsorption of Materials Such a
    Instrument Calibration (Flow Cytometry, Microscopy, HTS, HCS), Flow Testing (Microfluidics, Blood Flow, Water Flow, and Air Flow), Cell Biology Tracers (Cell Differentiation and Cell Tracing), Immunoassays (Agglutination Tests, ELISA, Particle Capture, and Contrast Reagents)
    Suspension
    1762 A2/SO4 (Sulfate Group)
    15 mL
    DVB-Sulfate
    0.1 μm
    2°C to 8°C
    White
    Polystyrene
    1 Each
    1.055g/cm3 at 20°C
    1.591 at 20°C
    15 mL
    0.1 μm
    Sugerencias de productos

    Sugerencias de productos

    Videos
    SDS
    Documentos

    Documentos

    Certificados
    Ofertas especiales

    Ofertas especiales

    For Research Use Only. Not for use in diagnostic procedures.