missing translation for 'onlineSavingsMsg'
Learn More

Clorhidrato de DL-fenilalanina metiléster, 98 %, Thermo Scientific Chemicals

Catalog Number p-7020710
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
5619-07-8
C10H14ClNO2
215.68
MFCD00066113
SWVMLNPDTIFDDY-UHFFFAOYNA-N
dl-phenylalanine methyl ester hydrochloride, methyl 2-amino-3-phenylpropanoate hydrochloride, h-dl-phe-ome.hcl, h-l-phe-ome hcl, phenylalanine methyl ester hydrochloride, l-phenylalanine methylester hydrochloride, l-methyl phenylalanate hydrochloride, s-phenylalanine methyl ester hydrochloride, acmc-209oxm, acmc-1bga3
3084017
methyl 2-amino-3-phenylpropanoate hydrochloride
Cl.COC(=O)C(N)CC1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
5619-07-8
C10H14ClNO2
215.68
MFCD00066113
SWVMLNPDTIFDDY-UHFFFAOYNA-N
dl-phenylalanine methyl ester hydrochloride, methyl 2-amino-3-phenylpropanoate hydrochloride, h-dl-phe-ome.hcl, h-l-phe-ome hcl, phenylalanine methyl ester hydrochloride, l-phenylalanine methylester hydrochloride, l-methyl phenylalanate hydrochloride, s-phenylalanine methyl ester hydrochloride, acmc-209oxm, acmc-1bga3
3084017
methyl 2-amino-3-phenylpropanoate hydrochloride
Cl.COC(=O)C(N)CC1=CC=CC=C1

DL-Phenylalanine methyl ester hydrochloride, is an aminoacid which is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El clorhidrato de DL-fenilalanina metiléster es un aminoácido que se utiliza ampliamente en productos farmacéuticos y alimentos.

Solubilidad
Ligeramente soluble en agua.

Notas
Almacenar en un lugar fresco. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado. Estable en las condiciones de almacenamiento recomendadas. Manténgase alejado de agentes oxidantes fuertes, bases fuertes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material DL-Phenylalanine methyl ester hydrochloride
Punto de fusión ∼160°C (decomposition)
Beilstein 3597947
Información de solubilidad Slightly soluble in water.
Formula Weight (peso de la fórmula) 215.68
Porcentaje de pureza 98%
Cantidad 5 g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'