missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANSELL™ Chemical and Mechanical Protection AlphaTec™ Gloves
Descripción
- Flock lining provides comfort to the hand and helps to improve moisture management
- Superior raised grip pattern for safety handling
- Resistance to many solvents and oil-based chemicals
Especificaciones
Certificaciones/conformidad (Europa) | CE 0493|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|EN 388:2016 +A1:2018|Food Contact|Category III|REACH Compliant|UKCA 0321 |
Chlorinated | Yes |
Aplicaciones | Industrial |
Color | Blue |
Finger Thickness (English) | 13.8 mil |
Finger Thickness (Metric) | 0.35 mm |
Latex Free | Yes |
Material | Nitrile |
Palm Thickness (English) | 13.8 mil |
Palm Thickness (Metric) | 0.35 mm |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'