Learn More
CENPV Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA560049
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: NLDLGEQRER WETFQKRQKL TSEGAAKLLL DTFEYQGLVK HTGGCHCGAV RFEVWASADL HIFDCNCSIC KKKQN Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 71%.
CENPV orthologs were detected in a range of species, including plants, but the proline-rich region was detected only in primates, rodents, birds, and amphibians, and long proline stretches were found only in primates. Northern blot analysis showed ubiquitous Cenpv expression in mouse tissues, with highest expression in testis, followed by small intestine, brain, and kidney. Immunofluorescence microscopy of mouse fibroblasts revealed colocalization of Cenpv with stabilized cytoplasmic microtubules. Western blot analysis of Ramos B cells detectedCENPV at an apparent molecular mass of 40 kD.Especificaciones
CENPV | |
Polyclonal | |
Unconjugated | |
CENPV | |
3110013H01Rik; AA671303; Cenpv; CENP-V; Centromere protein V; nuclear protein p30; p30; proline rich 6; proline-rich polypeptide 6; proline-rich protein 6; Prr6; RGD1565681 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z7K6 | |
CENPV | |
Recombinant protein corresponding to Human CENPV | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only