missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS45 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA518399
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
VPS45 Polyclonal antibody specifically detects VPS45 in Human samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
VPS45 | |
Polyclonal | |
Unconjugated | |
VPS45 | |
AI462172; AW554165; H1; H1VPS45; HGNC:14579; hlVps45; h-VPS45; leucocyte vacuolar protein sorting 45; mVps45; RP11-458I7.2; rVps45; SCN5; vacuolar protein sorting 45 (yeast); vacuolar protein sorting 45 homolog; vacuolar protein sorting 45 homolog (S. cerevisiae); vacuolar protein sorting 45A; vacuolar protein sorting protein 45; vacuolar protein sorting-associated protein 45; vesicular transport protein rvps45; Vps45; VPS45A; VPS45B; VPS54A; VSP45; Vsp45a | |
Goat | |
Antigen affinity chromatography | |
RUO | |
11311 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9NRW7 | |
VPS45 | |
Synthetic peptide sequence (FQKKKPKEQQKLES) corresponding to the internal amino acids of VPS45 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |