missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM106A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534352
Detalles adicionales : Peso : 0.01000kg
Descripción
A suggested positive control is A-20 cell lysate. PA5-34352 can be used with blocking peptide PEP-1395.
TMEM106A Polyclonal antibody specifically detects TMEM106A in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
TMEM106A | |
Polyclonal | |
Unconjugated | |
TMEM106A | |
0610008L10Rik; AI043106; BC022145; TMEM106A; Transmembrane protein 106A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113277, 217203 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8VC04, Q96A25 | |
TMEM106A | |
A 14 amino acid synthetic peptide near the amino terminus of human TMEM106A | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |