missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tankyrase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA132225
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PA1-32225 detects Tankyrase from human samples. PA1-32225 has been successfully used in Western blot applications. The PA1-32225 immunogen is: Synthetic peptide internal to tankyrase.
The approximately142 kDa Tankyrase protein is ubiquitously expressed in mammalian cells and is localized to human telomeres. It has homology to ankyrins and to the catalytic domain of poly (adenosine diphosphate-ribose) polymerase (PARP). Tankyrase binds to TRF1 (telomeric repeat binding factor-1), which negatively regulates the maintece of telomere length.Especificaciones
Tankyrase | |
Polyclonal | |
Unconjugated | |
O95271 | |
TNKS | |
Synthetic peptide internal to tankyrase. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1.1 mg/mL | |
TNKS | |
PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1, pART5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8658 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |