missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA540488
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: MDDAHESPSD KGGETGESDE TAAVPGDPGA TDTDGIPEET DGDADVDLKE Sequence homology: Cow: 92%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 83%; Rabbit: 92%; Rat: 100%
TAF11 Polyclonal antibody specifically detects TAF11 in Human samples. It is validated for Western Blot.Especificaciones
TAF11 | |
Polyclonal | |
Unconjugated | |
TAF11 | |
1110038O14Rik; 28kDa; AI115401; AL024207; AW552398; hypothetical protein LOC507047; MGC:15243; PRO2134; PRO214; TAF(II)28; Taf11; TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 28kDa; TAF2I; TAFII28; TAFII-28; TATA box binding protein (TBP)-associated factor 11; TATA box binding protein (TBP)-associated factor, RNA polymerase II, I, 28kD; TATA box binding protein associated factor 11; TATA-box binding protein associated factor 11; TBP-associated factor 11; TFIID subunit p30-beta; transcription initiation factor TFIID 28 kD subunit; transcription initiation factor TFIID 28 kDa subunit; Transcription initiation factor TFIID subunit 11 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6882 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15544 | |
TAF11 | |
Synthetic peptide directed towards the N-terminal of human TAF11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |