missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 1 Monoclonal Antibody (SP8), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA517616
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-17616 targets Syntaxin-1A in ELISA, FACS, and WB applications and shows reactivity with Human, Mouse, Porcine, and Rat samples. The MA5-17616 immunogen is human Syntaxin.
Syntaxin 1 is a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE [Soluble NSF (N-ethylmaleimide-sensitive fusion protein)-Attachment protein REceptor] domain (known as H3), and an N-terminal regulatory domain (Habc). Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. This gene product is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Especificaciones
Syntaxin 1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
O35526, P32851, Q16623 | |
Stx1a | |
Human Syntaxin | |
100 μg | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
SP8 | |
Biotin | |
Stx1a | |
HPC-1; LOW QUALITY PROTEIN: syntaxin-1A; neuron-specific antigen HPC-1; P35-1; P35A; Sap; STX1; Stx1a; STX1B; STX1B1; STX1B2; STXA1; SYN1A; Synaptotagmin-associated 35 kDa protein; syntaxin 1 a; syntaxin 1A; syntaxin 1A (brain); syntaxin 1A (brain)-like; syntaxin-1A; Syntaxin-1B1; Syntaxin-1B2 | |
Mouse | |
Protein G | |
RUO | |
100512594, 116470, 20907, 6804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |