missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAMF9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA521103
Detalles adicionales : Peso : 0.01000kg
Descripción
A suggested positive control is mouse kidney tissue lysate. PA5-21103 can be used with blocking peptide PEP-1217.
SLAMF9 Polyclonal antibody specifically detects SLAMF9 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
SLAMF9 | |
Polyclonal | |
Unconjugated | |
SLAMF9 | |
2310026I04Rik; AI462096; CD2 antigen family, member 10; CD2 family member 10; CD2F10; CD2F-10; CD84 homolog 1; CD84H1; CD84-H1; cluster of differentiation 2 antigen family member 10; cluster of differentiation 84 homolog 1; RP11-48O20.3; SF2001; signaling lymphocytic activation molecule family member 2001; signaling lymphocytic activation molecule family member 9; SLAM family member 9; SLAMF9; UNQ1938/PRO4421 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
289235, 89886, 98365 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q96A28, Q9D780 | |
SLAMF9 | |
A 19 amino acid peptide near the carboxy terminus of human SLAMF9. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |