Learn More
S100B Monoclonal Antibody (SH-B4), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA125005
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: human tongue. MA1-25005 has a predicted molecular weight of 10.5 kDa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
S100B Monoclonal antibody specifically detects S100B in Bovine, Canine, Feline, Goat, Human, Ovine, Porcine, Rabbit, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Especificaciones
S100B | |
Monoclonal | |
13.5 mg/mL | |
Ascites with 15mM sodium azide | |
P02638, P04271, P04631, Q6YNR6 | |
S100B | |
Purified, full length, native bovine S100b protein. | |
RUO | |
100009495, 100623510, 101099488, 101113190, 102169524, 25742, 491615, 525716, 6285 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
SH-B4 | |
Unconjugated | |
S100B | |
AI850290; Bpb; hypothetical protein LOC525716; NEF; protein S100-B; S100; S100 calcium binding protein B; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100B; S100-B; S100beta; S100P | |
Mouse | |
100 μL | |
Primary | |
Bovine, Canine, Feline, Goat, Human, Ovine, Porcine, Rabbit, Rat | |
Antibody | |
IgG1 |