Learn More
RT1.Ac Monoclonal Antibody (OX-27), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA517464
Detalles adicionales : Peso : 0.01000kg
Descripción
anti-rat RT1.Ac recognizes a polymorphic determinant of the MHC class I antigen in the rat. (“c” and “n” haplotypes). This antibody is an excellent antibody for labelling cells of donor or host origin in bone marrow chimeras. Applications include flow cytometry and immunohistochemistry using cryostat sections (however, cross-reactivity with Lewis rats have been shown to occur in some instances).
RT1.Ac Monoclonal antibody specifically detects RT1.Ac in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Especificaciones
RT1.Ac | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen) | |
OX-27 | |
Biotin | |
RT1-Ac | |
Phytohaemagglutinin Blasts | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |