missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RACK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA517484
Detalles adicionales : Peso : 0.01000kg
Descripción
It is not recommended to aliquot this antibody.
RACK1 Polyclonal antibody specifically detects RACK1 in Drosophila, Human, Mouse, Rat, Zebrafish samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
RACK1 | |
Polyclonal | |
Unconjugated | |
RACK1 | |
12-3; activated protein kinase C receptor; AL033335; Cell proliferation-inducing gene 21 protein; CG7111; CG7111-PA; CG7111-PB; CG7111-PC; CG7111-PD; Dmel\CG7111; Dmel_CG7111; dRack1; GB-like; gnb2l1; Gnb2-rs1; guanine nucleotide binding protein (G protein) beta polypeptide 2-like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1 sequence 1; guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1; guanine nucleotide binding protein beta polypeptide 2-like 1; guanine nucleotide binding protein related; guanine nucleotide binding protein, beta 2, related sequence 1; guanine nucleotide binding protein, beta polypeptide 2-like 1; guanine nucleotide binding protein, beta-2, related sequence 1; guanine nucleotide-binding protein beta-subunit like protein; guanine nucleotide-binding protein subunit beta-2-like 1; Guanine nucleotide-binding protein subunit beta-2-like 1, N-te | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10399, 14694, 30722, 34070, 83427 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
29 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
O18640, O42248, P63244, P63245, P68040 | |
RACK1 | |
Synthetic peptide corresponding to human RACK1 | |
100 μL | |
Primary | |
Drosophila, Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |