missing translation for 'onlineSavingsMsg'
Learn More

Progesterone Receptor Monoclonal Antibody (R.380.2), Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Invitrogen MA514841

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11530461

  • 410.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other PR family members.

Progesterone Receptor Monoclonal antibody specifically detects Progesterone Receptor in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

Progesterone Receptor
Monoclonal
40 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
P06401
PGR
Synthetic peptide corresponding to residues surrounding Tyr541 of human progesterone receptor
100 μL
Primary
Human
Antibody
IgG
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
R.380.2
Unconjugated
PGR
9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product
Rabbit
Affinity chromatography
RUO
5241
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales