missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP6C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA515607
Detalles adicionales : Peso : 0.01000kg
Descripción
PPP6C Polyclonal antibody specifically detects PPP6C in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
PPP6C | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O00743 | |
Ppp6c | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PPP6C | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Ppp6c | |
2310003C10Rik; ANKRD33; FLJ34236; PP6; PP6C; PPP6; PPP6C; Ppv; PP-V; protein phosphatase 6 catalytic subunit; protein phosphatase 6, catalytic subunit; protein phosphatase V; serine/threonine protein phosphatase catalytic subunit; serine/threonine-protein phosphatase 6 catalytic subunit; Serine/threonine-protein phosphatase 6 catalytic subunit, N-terminally processed; UNQ5837 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5537 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |