Learn More
PLSCR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA521649
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4. Predicted reactivity: Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLSCR1 Polyclonal antibody specifically detects PLSCR1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
PLSCR1 | |
Polyclonal | |
Unconjugated | |
PLSCR1 | |
ca(2+)-dependent phospholipid scramblase 1; Erythrocyte phospholipid scramblase; MmTRA1a; MmTRA1b; NOR1; Phospholipid scramblase 1; phospholipid scramblase 2; PL scramblase 1; Plscr1; Tra1; Tra1a; Tra1b; transplantability-associated protein 1; Tras1; Tras2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5359 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.52 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O15162 | |
PLSCR1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 286 of Scramblase1 (Uniprot ID#O15162) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |