missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IRS1 (Ser639) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA536828
Detalles adicionales : Peso : 0.00910kg
Descripción
This antibody detects endogenous protein at a molecular weight of 132 and 180 kDa. Purity is >95% by SDS-PAGE.
Phospho-IRS1 (Ser639) Polyclonal antibody specifically detects Phospho-IRS1 (Ser639) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
Phospho-IRS1 (Ser639) | |
Polyclonal | |
Unconjugated | |
IRS1 | |
G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16367, 25467, 3667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
P35568, P35569, P35570 | |
IRS1 | |
Synthetic phosphopeptide derived from human IRS-1 around the phosphorylation site of Serine 639 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |