Learn More
NUP35 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA531787
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (94%), Rat (95%), Zebrafish (80%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NUP35 Polyclonal antibody specifically detects NUP35 in Human samples. It is validated for Western Blot.Especificaciones
NUP35 | |
Polyclonal | |
Unconjugated | |
NUP35 | |
2310006I24Rik; 35 kDa nucleoporin; 35kDa; 5330402E05Rik; Mitotic phosphoprotein 44; Mp44; MP-44; NO44; NP44; Nuclear pore complex protein Nup53; nucleoporin 35; nucleoporin 35kDa; nucleoporin NUP35; Nucleoporin NUP53; Nup35; NUP53 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
129401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.87 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NFH5 | |
NUP35 | |
Recombinant fragment corresponding to a region within amino acids 127 and 326 of Human Nup53 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |