Learn More
MYL2 Monoclonal Antibody (7C9), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515496
Detalles adicionales : Peso : 0.01000kg
Descripción
MA5-15496 targets MYL2 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15496 immunogen is purified recombinant fragment of MYL2 expressed in E. Coli. <br>. MA5-15496 detects MYL2 which has a predicted molecular weight of approximately 19kDa.
MYL2 Monoclonal antibody specifically detects MYL2 in Human, Rat samples. It is validated for ELISA, Western Blot.Especificaciones
MYL2 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P08733, P10916 | |
MYL2 | |
Purified recombinant fragment of MYL2 expressed in E. Coli. | |
RUO | |
363925, 4633 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
7C9 | |
Unconjugated | |
MYL2 | |
cardiac myosin light chain 2; cardiac myosin light chain-2; cardiac ventricular myosin light chain 2; CMH10; DKFZp779C0562; hypothetical protein LOC677750; Mlc2; MLC-2; mLc2b; MLC-2s/v; MLC2V; MLC-2v; myl2; myl2b; Mylpc; myosin light chain 2; myosin light chain 2, slow skeletal/ventricular muscle isoform; myosin light chain 2V; myosin light chain phosphatase; myosin light chain, phosphorylatable, cardiac ventricles; myosin light chain-2; myosin regulatory light chain 2, ventricular/cardiac muscle isoform; myosin regulatory light chain 2b; myosin regulatory light chain ventricular isoform; myosin, light chain 2, regulatory, cardiac, slow; myosin, light chain 2b, regulatory, cardiac, slow; myosin, light polypeptide 2, regulatory, cardiac, slow; myosin, light polypeptide 2b, regulatory, cardiac, slow; regulatory light chain of myosin; RLC of myosin; slow cardiac myosin regulatory light chain 2; ventricular myosin light chain 2; ventricular myosin regulatory light chain; zgc:136848 | |
Mouse | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |