missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Monoclonal Antibody (22), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA125034
Detalles adicionales : Peso : 0.01000kg
Descripción
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MBP Monoclonal antibody specifically detects MBP in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Especificaciones
MBP | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
P02686 | |
MBP | |
Full length human protein. | |
RUO | |
4155 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
22 | |
Unconjugated | |
MBP | |
20 kDa microtubule-stabilizing protein; C76307; golli mbp; Golli-mbp; Golli-MBP; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; Hmbpr; MBP; MBP S; Mbps; MGC99675; microtubule-stabilizing protein; mLd; MOBP; Myelin A1 protein; myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; myelin membrane encephalitogenic protein; Myelin P1 protein; myelin-associated oligodendrocyte basic protein; R75289; shi; shiverer; Unknown (protein for IMAGE:7984318); unnamed protein product | |
Mouse | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b |